Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diclofenac Sodium, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
15307-79-6
C14H10Cl2NNaO2
318.13
KPHWPUGNDIVLNH-UHFFFAOYSA-M
sodium 2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetate
[Na+].[O-]C(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Cl
15307-79-6
C14H10Cl2NNaO2
318.13
KPHWPUGNDIVLNH-UHFFFAOYSA-M
sodium 2-{2-[(2,6-dichlorophenyl)amino]phenyl}acetate
[Na+].[O-]C(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Cl

D1287, 15307-79-6, C14H10Cl2NNaO2

Spectrum™ Diclofenac Sodium, USP is a nonsteroidal anti-inflammatory drug used to reduce inflammation and is as an analgesic. All Spectrum Chemical USP products are manufactured, packa ged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.005
Quantity 100 g
CAS Max % 1
UN Number UN2811
Percent Purity 99 to 101%