Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclohexano-24-crown-8, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
17455-23-1
C24H44O8
460.608
MFCD00005102
QMLGNDFKJAFKGZ-UHFFFAOYSA-N
dicyclohexano-24-crown-8,ccris 3621,dicyclohexyl 24-crown-8,dibenz b,n 1,4,7,10,13,16,19,22 octaoxacyclotetracosin, tetracosahydro,tetracosahydrodibenz b,n 1,4,7,10,13,16,19,22 octaoxacyclotetracosin,dicyclohexyl-24-crown-8,dicyclohexano-24-crown-8 ether,tetracosahydrodibenzo b,n 1,4,7,10,13,16,19,22 octaoxacyclotetracosine,2,5,8,11,18,21,24,27-octaoxatricyclo 26.4.0.0<12,17> dotriacontane
87119
C1CCC2C(C1)OCCOCCOCCOC3CCCCC3OCCOCCOCCO2
17455-23-1
C24H44O8
460.608
MFCD00005102
QMLGNDFKJAFKGZ-UHFFFAOYSA-N
dicyclohexano-24-crown-8,ccris 3621,dicyclohexyl 24-crown-8,dibenz b,n 1,4,7,10,13,16,19,22 octaoxacyclotetracosin, tetracosahydro,tetracosahydrodibenz b,n 1,4,7,10,13,16,19,22 octaoxacyclotetracosin,dicyclohexyl-24-crown-8,dicyclohexano-24-crown-8 ether,tetracosahydrodibenzo b,n 1,4,7,10,13,16,19,22 octaoxacyclotetracosine,2,5,8,11,18,21,24,27-octaoxatricyclo 26.4.0.0<12,17> dotriacontane
87119
C1CCC2C(C1)OCCOCCOCCOC3CCCCC3OCCOCCOCCO2
Chelation/Complexation reagent
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Color Yellow
Density 1.1020g/mL
Refractive Index 1.4875 to 1.4895 (20°C, 589nm)
Assay Percent Range 97%
Merck Index 14, 2602
Formula Weight 460.6
Physical Form Viscous Liquid
Percent Purity ≥96.0%
Chemical Name or Material Dicyclohexano-24-crown-8, 97%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Toxic if inhaled.
Causes serious eye irritation.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger