Learn More
Dicyclohexylamine, 98%, Thermo Scientific Chemicals
Description
Dicyclohexylamine is used as an intermediate for the production of antioxidants and vulcanization accelerator. It is used as a catalyst for flexible polyurethane foams. It is also used in agrochemicals and textile chemicals. Used to constitute ionic liquid matrices for bacterial analysis in matrix assisted laser desorption/ionisation mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dicyclohexylamine is used as an intermediate for the production of antioxidants and vulcanization accelerator. It is used as a catalyst for flexible polyurethane foams. It is also used in agrochemicals and textile chemicals. Used to constitute ionic liquid matrices for bacterial analysis in matrix assisted laser desorption/ionisation mass spectrometry.
Solubility
Sparingly soluble in water (0.8 g/L).
Notes
Air sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container.
Specifications
Melting Point | -2°C |
Density | 0.913 |
Boiling Point | 256°C |
Flash Point | 99°C (210°F) |
Refractive Index | 1.4842 |
Quantity | 500 mL |
UN Number | UN2565 |
Beilstein | 605923 |
Sensitivity | Air Sensitive |
Merck Index | 14,3095 |
Show More |
Safety and Handling
Hazard Category | H301+H311-H314-H335 |
Hazard Statement | GHS H Statement H314-H302 Causes severe skin burns and eye damage. Harmful if swallowed. |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: DICYCLOHEXYLAMINE |
EINECSNumber | 202-980-7 |
RTECSNumber | HY4025000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.