Alfa Aesar™ Dicyclohexylmethane 4,4'-diisocyanate, mixture of isomers, tech. 90%

Manufacturer: Alfa Aesar™ 44087AK

Quantity 250mL

 View more versions of this product

Catalog No. AA44087AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5124-30-1
    Molecular Formula C15H22N2O2
    Formula Weight 262.37
    MDL Number MFCD00015405
    UN Number UN2206
    Synonym 4,4'-Methylenebis(cyclohexyl isocyanate)
    Chemical Name or Material Dicyclohexylmethane 4,4'-diisocyanate, Mixture of isomers
    Grade Technical Grade

    Specifications

    Formula Weight 262.37
    Health Hazard 2 GHS H Statement
    H331-H334-H315-H319-H317-H335
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00015405
    Physical Form Liquid
    CAS 5124-30-1
    UN Number UN2206
    Boiling Point 168°C (1.5mmHg)
    Health Hazard 1 Danger
    Synonym 4,4'-Methylenebis(cyclohexyl isocyanate)
    Density 1.066g/mL
    RTECS Number NQ9250000
    Flash Point >110°C (230°F)
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
    TSCA TSCA
    Molecular Formula C15H22N2O2
    Quantity 250mL
    Chemical Name or Material Dicyclohexylmethane 4,4'-diisocyanate, Mixture of isomers
    Assay 90%
    Grade Technical Grade

    GHS H Statement
    H331-H334-H315-H319-H317-H335
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P284a-P261-P280-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit