Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dicyclohexylphenylphosphine, 98%

Manufacturer:  Alfa Aesar™ H2699203

Quantity 1g

 View more versions of this product

Catalog No. AAH2699203


  Check Availability
Add to cart

Specifications

Description

Applications
Dicyclohexylphenylphosphine, is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
Description & Specifications

Chemical Identifiers

6476-37-5
274.38
Dicyclohexylphenylphosphine
C18H27P
MFCD00003854

Specifications

1g
C18H27P
MFCD00003854
Dicyclohexylphenylphosphine
6476-37-5
274.38
58° to 61°C
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning

RTECSNumber : SY9125000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit