Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclopentadiene diepoxide, 98%, Acros Organics™

Click to view available options
Quantity:
100g
81-21-0
C10H12O2
164.2
BQQUFAMSJAKLNB-UHFFFAOYSA-N
dicyclopentadiene dioxide,dicyclopentadiene diepoxide,unox epoxide 207,epoxide 207,bicyclopentadiene dioxide,unox 207x,unox 207,dicyclopentadiene dioxide van,1,2:5,6-diepoxyhexahydro-4,7-methanoindan,4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
6673
C1C2C3CC4C(C3C1C5C2O5)O4
81-21-0
C10H12O2
164.2
BQQUFAMSJAKLNB-UHFFFAOYSA-N
dicyclopentadiene dioxide,dicyclopentadiene diepoxide,unox epoxide 207,epoxide 207,bicyclopentadiene dioxide,unox 207x,unox 207,dicyclopentadiene dioxide van,1,2:5,6-diepoxyhexahydro-4,7-methanoindan,4,7-methanoindan, 1,2:5,6-diepoxyhexahydro
6673
C1C2C3CC4C(C3C1C5C2O5)O4
Quantity 100g
Infrared Spectrum Authentic
Melting Point 185.0°C to 189.0°C
Boiling Point 326.0°C
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: 1.4g/L (20°C).
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material Dicyclopentadiene diepoxide

GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove

GHS Signal Word: Danger

EINECSNumber : 201-334-1