Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclopentadiene, Stabilized 95%, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
77-73-6,98-29-3
C10H12
132.2
HECLRDQVFMWTQS-UHFFFAOYSA-N
dicyclopentadiene,biscyclopentadiene,cyclopentadiene dimer,bicyclopentadiene,dicyklopentadien,dimer cyklopentadienu,dcpd,4,7-methano-1h-indene, 3a,4,7,7a-tetrahydro,3a,4,7,7a-tetrahydro-1h-4,7-methanoindene,1,3-cyclopentadiene, dimer
6492
CHEBI:34695
C1C=CC2C1C3CC2C=C3
77-73-6,98-29-3
C10H12
132.2
HECLRDQVFMWTQS-UHFFFAOYSA-N
dicyclopentadiene,biscyclopentadiene,cyclopentadiene dimer,bicyclopentadiene,dicyklopentadien,dimer cyklopentadienu,dcpd,4,7-methano-1h-indene, 3a,4,7,7a-tetrahydro,3a,4,7,7a-tetrahydro-1h-4,7-methanoindene,1,3-cyclopentadiene, dimer
6492
CHEBI:34695
C1C=CC2C1C3CC2C=C3
Specific Gravity 0.982
Stabilizer 100 to 200ppm p-tert.-butylcatechol
Quantity 10L
Refractive Index 1.5100 to 1.5120
Boiling Point 170.0°C
Water 0.03% Max. (K.F.)
Color Undesignated
Melting Point 11.0°C to 13.0°C
Flash Point 32°C
Packaging Plastic drum
Density 0.9820g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Beilstein 05,II,391
Merck Index 14,2739
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ethanol,diethyl ether
Formula Weight 132.2
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Dicyclopentadiene
Show More Show Less