Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Didodecylamine 97.0+%, TCI America™
SDP

Catalog No. p-7126114
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 3007-31-6
Molecular Formula C24H51N
Molecular Weight (g/mol) 353.679
MDL Number MFCD00041918
InChI Key MJCJUDJQDGGKOX-UHFFFAOYSA-N
Synonym didodecylamine, di-n-dodecylamine, 1-dodecanamine, n-dodecyl, dilaurylamine, alamine 204, n-dodecyl-1-dodecanamine, unii-5kr2ml6h0c, 5kr2ml6h0c, dilauryl amine, didodecyl amine
PubChem CID 18157
IUPAC Name N-dodecyldodecan-1-amine
SMILES CCCCCCCCCCCCNCCCCCCCCCCCC

Specifications

Melting Point 51°C
Color White-Yellow
Boiling Point 265°C
Quantity 5 g
Formula Weight 353.68
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Didodecylamine
EINECSNumber (2)-0176&(2)-0138
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.