Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diethanolamine, ≥99.7% (GC), MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 169571MLF
Description
Diethanolamine (DEA) is an organic chemical compound, which comprises of a secondary amine and a dialcohol. It mainly acts as a weak base. DEA can be used in preparing long-chain fatty acids, namely diethanolamides and diethanolamine salts, which find application in liquid laundry and dishwashing detergents, cosmetics, shampoos, hair conditioners, textile processing, and also in pharmaceutical industries. Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal CareSpecifications
111-42-2 | |
Analytical Standard | |
MFCD00002843 | |
105.14 | |
Analytical Standard | |
<0.98 atm (100°C) | |
1.097 g/mL (at 25°C) | |
217°C (150 mmHg) | |
Neat |
1 mL | |
HN(CH2 CH2 OH)2 | |
2,2′-Iminodiethanol; Bis(2-hydroxyethyl)amine | |
≥99.7% (GC) | |
689°F | |
3.6 (vs. air) | |
28°C | |
138°C |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction