Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethanolamine, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

111-42-2
C4H11NO2
105.137
ZBCBWPMODOFKDW-UHFFFAOYSA-N
8113
CHEBI:28123
2-(2-hydroxyethylamino)ethanol
C(CO)NCCO

High quality chemicals for laboratory and specialized industrial use

Packaging Narrow Mouth Amber Glass Bottle
Quantity 4L
Flash Point 130.00°C
Specific Gravity 1L = 1.09kg
Formula Weight 105.14
Chemical Name or Material Diethanolamine
Grade Baker Analyzed™ ACS Reagent