Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethanolamine, Reagent, ACS, 98.5%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
111-42-2
C4H11NO2
105.14
ZBCBWPMODOFKDW-UHFFFAOYSA-N
2-[(2-hydroxyethyl)amino]ethan-1-ol
OCCNCCO
111-42-2
C4H11NO2
105.14
ZBCBWPMODOFKDW-UHFFFAOYSA-N
2-[(2-hydroxyethyl)amino]ethan-1-ol
OCCNCCO

D1033, 111-42-2, C4H11NO2

Spectrum™ Diethanolamine, Reagent, ACS is also referred to as DEA, is an organic compound that in its pure form is a white solid but it is most often encountered as a clear liquid that has a viscous texture. It is a secondary amine and diol and is water soluble. It is used as a corrosion inhibitor and surfactant that is used in the oil refining industry to help remove hydrogen sulfide from sour gas. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.
Packaging Steel Can
Water 0.0015
Quantity 20 L
CAS Max % 1
Percent Purity 98.5%