Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethoxydimethylsilane, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
250mL
500mL
78-62-6
C6H16O2Si
148.277
MFCD00009068
YYLGKUPAFFKGRQ-UHFFFAOYSA-N
dimethyldiethoxysilane,silane, diethoxydimethyl,diethoxy dimethyl silane,dimethyl-diethoxysilan,dimethylsilicondiethoxide,dimethyl diethoxy silane,unii-i3931j3zj2,dimethyl-diethoxysilan czech,ccris 1321,kbe 22
62322
diethoxy(dimethyl)silane
CCO[Si](C)(C)OCC
78-62-6
C6H16O2Si
148.277
MFCD00009068
YYLGKUPAFFKGRQ-UHFFFAOYSA-N
dimethyldiethoxysilane,silane, diethoxydimethyl,diethoxy dimethyl silane,dimethyl-diethoxysilan,dimethylsilicondiethoxide,dimethyl diethoxy silane,unii-i3931j3zj2,dimethyl-diethoxysilan czech,ccris 1321,kbe 22
62322
diethoxy(dimethyl)silane
CCO[Si](C)(C)OCC
Infrared Spectrum Authentic
Density 0.86g/mL
Quantity 25mL
Refractive Index 1.3805 to 1.3825 (20°C, 589nm)
Color Colorless
Assay Percent Range 97%
Linear Formula (CH3)2Si(OC2H5)2
Fieser 13,76
Formula Weight 148.28
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Diethoxydimethylsilane, 97%
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger