Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethoxydimethylsilane, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
250mL
500mL
78-62-6
C6H16O2Si
148.28
MFCD00009068
YYLGKUPAFFKGRQ-UHFFFAOYSA-N
dimethyldiethoxysilane,silane, diethoxydimethyl,diethoxy dimethyl silane,dimethyl-diethoxysilan,dimethylsilicondiethoxide,dimethyl diethoxy silane,unii-i3931j3zj2,dimethyl-diethoxysilan czech,ccris 1321,kbe 22
62322
diethoxy(dimethyl)silane
CCO[Si](C)(C)OCC
78-62-6
C6H16O2Si
148.28
MFCD00009068
YYLGKUPAFFKGRQ-UHFFFAOYSA-N
dimethyldiethoxysilane,silane, diethoxydimethyl,diethoxy dimethyl silane,dimethyl-diethoxysilan,dimethylsilicondiethoxide,dimethyl diethoxy silane,unii-i3931j3zj2,dimethyl-diethoxysilan czech,ccris 1321,kbe 22
62322
diethoxy(dimethyl)silane
CCO[Si](C)(C)OCC
Specific Gravity 0.86
Infrared Spectrum Authentic
Color Colorless
Packaging Glass bottle
Melting Point -70.0°C
Quantity 100mL
Boiling Point 114.0°C
Refractive Index 1.3805 to 1.3825
Density 0.8600g/mL
Flash Point 11°C
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)2Si(OC2H5)2
Fieser 13,76
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in most common organic solvents
Formula Weight 148.28
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Diethoxydimethylsilane, 97%
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautio

GHS Signal Word: Danger

EINECSNumber : 201-127-6