Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethoxydiphenylsilane, 98%

Click to view available options
Quantity:
10g
50g
250g
2553-19-7
C16H20O2Si
272.419
MFCD00015126
ZZNQQQWFKKTOSD-UHFFFAOYSA-N
diphenyldiethoxysilane,silane, diethoxydiphenyl,diethoxy diphenyl silane,benzene, 1,1'-diethoxysilylene bis,acmc-1cmgu,diethoxy-di phenyl silane,diethoxydiphenylsilane,zznqqqwfkktosd-uhfffaoysa
75705
diethoxy(diphenyl)silane
CCO[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OCC
2553-19-7
C16H20O2Si
272.419
MFCD00015126
ZZNQQQWFKKTOSD-UHFFFAOYSA-N
diphenyldiethoxysilane,silane, diethoxydiphenyl,diethoxy diphenyl silane,benzene, 1,1'-diethoxysilylene bis,acmc-1cmgu,diethoxy-di phenyl silane,diethoxydiphenylsilane,zznqqqwfkktosd-uhfffaoysa
75705
diethoxy(diphenyl)silane
CCO[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OCC
Beilstein 2281302
Formula Weight 272.42
Percent Purity 98%
Boiling Point 166°C to 168°C (15mmHg)
Quantity 250g
Flash Point 175°C (347°F)
Chemical Name or Material Diethoxydiphenylsilane

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA