Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethoxymethane, 99%, stab. with 100ppm BHT, Alfa Aesar™

Click to view available options
Quantity:
25mL
100mL
500mL
462-95-3
C5H12O2
104.149
MFCD00009246
KLKFAASOGCDTDT-UHFFFAOYSA-N
diethoxymethane,ethylal,formaldehyde diethyl acetal,diethylformal,diethoxy methane,methane, diethoxy,ethoxymethoxy ethane,ethoxymethyl ethyl ether,1,1-diethoxymethane,3,5-dioxaheptene
10024
ethoxymethoxyethane
CCOCOCC
462-95-3
C5H12O2
104.149
MFCD00009246
KLKFAASOGCDTDT-UHFFFAOYSA-N
diethoxymethane,ethylal,formaldehyde diethyl acetal,diethylformal,diethoxy methane,methane, diethoxy,ethoxymethoxy ethane,ethoxymethyl ethyl ether,1,1-diethoxymethane,3,5-dioxaheptene
10024
ethoxymethoxyethane
CCOCOCC
Quantity 500mL
Melting Point -66°C
Flash Point −5°C (23°F)
Boiling Point 87°C to 88°C
Assay Percent Range 0.99
UN Number UN2373
Beilstein 1697253
Formula Weight 104.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Diethoxymethane, stabilized with 100ppm BHT
Show More Show Less

GHS H Statement
H225-H312-H315-H319-H335
Highly flammable liquid and vapour.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIETHOXYMETHANE

RTECSNumber : PA8500000

TSCA : TSCA