Alfa Aesar™ Diethoxymethane, 99%, stab. with 100ppm BHT

Manufacturer: Alfa Aesar™ B23134AC

Quantity 25mL

 View more versions of this product

Catalog No. AAB23134AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 462-95-3
    Molecular Formula CH3CH2OCH2OCH2CH3
    Formula Weight 104.15
    MDL Number MFCD00009246
    UN Number UN2373
    Beilstein 1697253
    Synonym Ethylal; Formaldehyde diethyl acetal
    Chemical Name or Material Diethoxymethane, stabilized with 100ppm BHT

    Specifications

    Formula Weight 104.15
    Physical Form Liquid
    Assay Percent Range 0.99
    Molecular Formula CH3CH2OCH2OCH2CH3
    MDL Number MFCD00009246
    UN Number UN2373
    Boiling Point 87° to 88°C
    Beilstein 1697253
    Melting Point −66°C
    Flash Point −5°C (23°F)
    Health Hazard 1 Danger
    Quantity 25mL
    TSCA TSCA
    CAS 462-95-3
    Synonym Ethylal; Formaldehyde diethyl acetal
    Health Hazard 2 GHS H Statement
    H225-H312-H315-H319-H335
    Highly flammable liquid and vapour.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIETHOXYMETHANE
    RTECS Number PA8500000
    Chemical Name or Material Diethoxymethane, stabilized with 100ppm BHT
    Assay 99%

    GHS H Statement
    H225-H312-H315-H319-H335
    Highly flammable liquid and vapour.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit