Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethoxymethyleacetate, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
14036-06-7
C7H14O4
162.185
IRUNKQSGDBYUDC-UHFFFAOYSA-N
methanol, diethoxy-, acetate,diethoxymethylacetate,acetic acid diethoxymethyl ester,methanol, 1,1-diethoxy-, 1-acetate,methyl2,2-diethoxyacetate,pubchem13756,acmc-209ckk,diethoxymethyl acetate,qspl 099,diethoxy-acetic acid methyl ester
84166
diethoxymethyl acetate
CCOC(OCC)OC(=O)C
14036-06-7
C7H14O4
162.185
IRUNKQSGDBYUDC-UHFFFAOYSA-N
methanol, diethoxy-, acetate,diethoxymethylacetate,acetic acid diethoxymethyl ester,methanol, 1,1-diethoxy-, 1-acetate,methyl2,2-diethoxyacetate,pubchem13756,acmc-209ckk,diethoxymethyl acetate,qspl 099,diethoxy-acetic acid methyl ester
84166
diethoxymethyl acetate
CCOC(OCC)OC(=O)C
Color Colorless
Infrared Spectrum Authentic
Density 0.9900g/mL
Packaging Glass bottle
Flash Point 27°C
Refractive Index 1.3980 to 1.4000 (20°C, 589nm)
Quantity 5g
Assay Percent Range 96%
Linear Formula CH3CO2CH(OC2H5)2
Formula Weight 162.19
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Diethoxymethyl acetate
Show More Show Less

GHS H Statement:
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning