Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethoxymethylphenylsilane, 97+%

Click to view available options
Quantity:
10g
50g
775-56-4
C11H18O2Si
210.348
MFCD00048541
MNFGEHQPOWJJBH-UHFFFAOYSA-N
methylphenyldiethoxysilane,diethoxy methyl phenylsilane,diethoxy methyl phenyl silane,silane, diethoxymethylphenyl,benzene, diethoxymethylsilyl,pubchem10811,acmc-209pap,methyl-phenyl-diethoxysilane,diethoxy-methyl-phenyl-silane
69891
diethoxy-methyl-phenylsilane
CCO[Si](C)(C1=CC=CC=C1)OCC
775-56-4
C11H18O2Si
210.348
MFCD00048541
MNFGEHQPOWJJBH-UHFFFAOYSA-N
methylphenyldiethoxysilane,diethoxy methyl phenylsilane,diethoxy methyl phenyl silane,silane, diethoxymethylphenyl,benzene, diethoxymethylsilyl,pubchem10811,acmc-209pap,methyl-phenyl-diethoxysilane,diethoxy-methyl-phenyl-silane
69891
diethoxy-methyl-phenylsilane
CCO[Si](C)(C1=CC=CC=C1)OCC
Beilstein 2938396
Formula Weight 210.35
Percent Purity ≥97%
Boiling Point 117°C to 118°C (30mmHg)
Quantity 10g
Flash Point 88°C (190°F)
Chemical Name or Material Diethoxymethylphenylsilane

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA