Learn More
Diethyl 1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate, 98%, Thermo Scientific Chemicals
Description
Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is a hydrogen source in conjugate reduction and organocatalytic reductive amination. It is used as a hydrogen source in organocatalytic reductive amination and conjugate reduction.
Solubility
Soluble in organic solvents.
Notes
Desiccate at 4°C. Keep away from oxidizing agents.
Specifications
Melting Point | 178°C to 183°C |
Quantity | 5 g |
Solubility Information | Soluble in organic solvents. |
Formula Weight | 253.3 |
Percent Purity | 98% |
Chemical Name or Material | Diethyl 1,4-dihydro-2,6-dimethylpyridine-3,5-dicarboxylate |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 214-561-6 |
RTECSNumber | US7970100 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.