Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl 1-butylphosphonate, 99+%

Manufacturer:  Alfa Aesar™ B2399706

Quantity 5g

 View more versions of this product

Catalog No. AAB2399706


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2404-75-3
194.21
1763851
Diethyl 1-butylphosphonate
C8H19O3P
MFCD00082700
1-Butylphosphonic acid diethyl ester; Diethyl 1-butanephosphonate

Specifications

5g
Odorless
194.21
74° to 76°C (1mmHg)
1-Butylphosphonic acid diethyl ester; Diethyl 1-butanephosphonate
≥99%
2404-75-3
C8H19O3P
MFCD00082700
1763851
Diethyl 1-butylphosphonate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit