Alfa Aesar™ Diethyl 1-butylphosphonate, 99+%

Manufacturer: Alfa Aesar™ B2399714

Quantity 25g

 View more versions of this product

Catalog No. AAB2399714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2404-75-3
    Molecular Formula C8H19O3P
    Formula Weight 194.21
    MDL Number MFCD00082700
    Beilstein 1763851
    Synonym 1-Butylphosphonic acid diethyl ester; Diethyl 1-butanephosphonate
    Chemical Name or Material Diethyl 1-butylphosphonate

    Specifications

    Formula Weight 194.21
    Boiling Point 74° to 76°C (1mmHg)
    Quantity 25g
    Odor Odorless
    Molecular Formula C8H19O3P
    MDL Number MFCD00082700
    CAS 2404-75-3
    Beilstein 1763851
    Synonym 1-Butylphosphonic acid diethyl ester; Diethyl 1-butanephosphonate
    Chemical Name or Material Diethyl 1-butylphosphonate
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit