Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl 1-butylphosphonate, 99+%

Click to view available options
Quantity:
5g
25g
100g
2404-75-3
C8H19O3P
194.211
MFCD00082700
KEOJOAVYGLGAIM-UHFFFAOYSA-N
diethyl butylphosphonate,diethyl 1-butylphosphonate,phosphonic acid, butyl-, diethyl ester,diethyl n-butanephosphonate,butylphosphonic acid diethyl ester,phosphonic acid,p-butyl-, diethyl ester
2773687
1-diethoxyphosphorylbutane
CCCCP(=O)(OCC)OCC
2404-75-3
C8H19O3P
194.211
MFCD00082700
KEOJOAVYGLGAIM-UHFFFAOYSA-N
diethyl butylphosphonate,diethyl 1-butylphosphonate,phosphonic acid, butyl-, diethyl ester,diethyl n-butanephosphonate,butylphosphonic acid diethyl ester,phosphonic acid,p-butyl-, diethyl ester
2773687
1-diethoxyphosphorylbutane
CCCCP(=O)(OCC)OCC
Odor Odorless
Boiling Point 74°C to 76°C (1mmHg)
Quantity 100g
Beilstein 1763851
Formula Weight 194.21
Percent Purity ≥99%
Chemical Name or Material Diethyl 1-butylphosphonate