Alfa Aesar™ Diethyl 2,2-diethoxyethylphosphonate, 96%

Manufacturer: Alfa Aesar™ A1246118

Quantity 50g

 View more versions of this product

Catalog No. AAA1246118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7598-61-0
    Molecular Formula C10H23O5P
    Formula Weight 254.26
    MDL Number MFCD00009236
    Beilstein 1785614
    Synonym 2,2-Diethoxyethylphosphonic acid diethyl ester; Diethyl phosphonoacetaldehyde diethyl acetal
    Chemical Name or Material Diethyl 2,2-diethoxyethylphosphonate

    Specifications

    Formula Weight 254.26
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Molecular Formula C10H23O5P
    CAS 7598-61-0
    Flash Point >110°C (230°F)
    Recommended Storage 2°C to 8°C
    MDL Number MFCD00009236
    Beilstein 1785614
    Synonym 2,2-Diethoxyethylphosphonic acid diethyl ester; Diethyl phosphonoacetaldehyde diethyl acetal
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Quantity 50g
    Boiling Point 101° to 103°C (8mmHg)
    Chemical Name or Material Diethyl 2,2-diethoxyethylphosphonate
    Assay 96%

    Description

    Applications
    Diethyl 2,2-diethoxyethylphosphonate is used reactant for synthesis of α,β-alkenal derivatives by two-carbon homologation, lower rim-phosphonylated rexorcinol calix[4]arenes by condensation reactions, α-Phosphovinyl radicals via a radical trapping sequence, inhibitors of reverse transcriptase via 1,3-dipolar cycloadditions and for Friedel-Crafts reactions.

    Solubility
    Soluble in chloroform and dichloromethane. Not miscible or difficult to mix in water.

    Notes
    Keep container tightly sealed. It is hygroscopic in nature. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    2°C to 8°C

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit