Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Diethyl 2,3-O-isopropylidene-L-tartrate, 97%, Thermo Scientific™

Catalog Number ac277570025
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
59779-75-8
C11H18O6
246.26
MFCD00143372
MCNKHXZIPATURB-HTQZYQBOSA-N
4r,5r-diethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, diethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4r,5r-4,5-diethoxycarbonyl-2,2-dimethyl dioxolane, 2-o,3-o-isopropylidene-l-tartaric acid diethyl ester, 4r,5r-4,5-bis ethoxycarbonyl-2,2-dimethyl-1,3-dioxolane, 4,5-diethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 1,3-dioxolane-4,5-dicarboxylicacid, 2,2-dimethyl-, 4,5-diethyl ester, 4r,5r, ethyl 4r,5r-5-ethoxycarbonyl-2,2-dimethyl-1,3-dioxolane-4-carboxylate
774789
diethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
CCOC(=O)C1C(OC(O1)(C)C)C(=O)OCC
59779-75-8
C11H18O6
246.26
MFCD00143372
MCNKHXZIPATURB-HTQZYQBOSA-N
4r,5r-diethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, diethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4r,5r-4,5-diethoxycarbonyl-2,2-dimethyl dioxolane, 2-o,3-o-isopropylidene-l-tartaric acid diethyl ester, 4r,5r-4,5-bis ethoxycarbonyl-2,2-dimethyl-1,3-dioxolane, 4,5-diethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 1,3-dioxolane-4,5-dicarboxylicacid, 2,2-dimethyl-, 4,5-diethyl ester, 4r,5r, ethyl 4r,5r-5-ethoxycarbonyl-2,2-dimethyl-1,3-dioxolane-4-carboxylate
774789
diethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
CCOC(=O)C1C(OC(O1)(C)C)C(=O)OCC
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Boiling Point 92.0°C to 93.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4345 to 1.4365
Quantity 2.5 g
Formula Weight 246.26
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (-)-Diethyl 2, 3-O-isopropylidene-L-tartrate, 97%
Show More Show Less

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit