Alfa Aesar™ Diethyl 2-acetylglutarate, 98+%

Manufacturer: Alfa Aesar™ B2445622

Quantity 100g

 View more versions of this product

Catalog No. AAB2445622

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1501-06-0
    Molecular Formula C11H18O5
    Formula Weight 230.27
    MDL Number MFCD00009158
    Beilstein 1371947
    Synonym 2-Acetylglutaric acid diethyl ester
    Chemical Name or Material Diethyl 2-acetylglutarate

    Specifications

    Formula Weight 230.27
    Molecular Formula C11H18O5
    MDL Number MFCD00009158
    CAS 1501-06-0
    Boiling Point 181° to 183°C (50mmHg)
    Beilstein 1371947
    Flash Point >110°C (230°F)
    Synonym 2-Acetylglutaric acid diethyl ester
    Quantity 100g
    Chemical Name or Material Diethyl 2-acetylglutarate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit