Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl (2-pentyl)malonate, 94%

Click to view available options
Quantity:
10g
50g
250g
117-47-5
C12H22O4
230.304
MFCD00051558
RQFSNEWORATSCC-UHFFFAOYSA-N
diethyl 1-methylbutyl malonate,diethyl 2-pentyl malonate,diethyl 1-methylbutylmalonate,diethyl 1-methylbutal malonate,1-methylbutyl malonic ester,1-methylbutyl malonic acid diethyl ester,diethyl pentan-2-ylmalonate,acmc-1brsl,diethyl sec-amyl malonate
95449
diethyl 2-pentan-2-ylpropanedioate
CCCC(C)C(C(=O)OCC)C(=O)OCC
117-47-5
C12H22O4
230.304
MFCD00051558
RQFSNEWORATSCC-UHFFFAOYSA-N
diethyl 1-methylbutyl malonate,diethyl 2-pentyl malonate,diethyl 1-methylbutylmalonate,diethyl 1-methylbutal malonate,1-methylbutyl malonic ester,1-methylbutyl malonic acid diethyl ester,diethyl pentan-2-ylmalonate,acmc-1brsl,diethyl sec-amyl malonate
95449
diethyl 2-pentan-2-ylpropanedioate
CCCC(C)C(C(=O)OCC)C(=O)OCC
Applications
Diethyl (2-pentyl)malonate is used in vitro reaction of barbiturates with formaldehyde to evaluate forensic toxicology on the cause of death.

Notes
Incompatible with oxidizing agents.
Quantity 10g
Boiling Point 118°C to 119°C (9mmHg)
Formula Weight 230.31
Percent Purity 94%
Chemical Name or Material Diethyl (2-pentyl)malonate