Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Diethyl (2-pentyl)malonate, 94%
Click to view available options
Quantity:
10g
50g
250g
Description
Applications
Diethyl (2-pentyl)malonate is used in vitro reaction of barbiturates with formaldehyde to evaluate forensic toxicology on the cause of death.
Notes
Incompatible with oxidizing agents.
Diethyl (2-pentyl)malonate is used in vitro reaction of barbiturates with formaldehyde to evaluate forensic toxicology on the cause of death.
Notes
Incompatible with oxidizing agents.
Specifications
Refractive Index | 1.427 |
Density | 0.968 |
Boiling Point | 118°C to 119°C (9mmHg) |
Quantity | 50g |
Formula Weight | 230.31 |
Percent Purity | 94% |
Chemical Name or Material | Diethyl (2-pentyl)malonate |
Safety and Handling
EINECSNumber : 204-193-4
TSCA : No
Recommended Storage : Ambient temperatures