Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl (2-pentyl)malonate, 94%

Click to view available options
Quantity:
10g
50g
250g
117-47-5
C12H22O4
230.304
MFCD00051558
RQFSNEWORATSCC-UHFFFAOYSA-N
diethyl 1-methylbutyl malonate,diethyl 2-pentyl malonate,diethyl 1-methylbutylmalonate,diethyl 1-methylbutal malonate,1-methylbutyl malonic ester,1-methylbutyl malonic acid diethyl ester,diethyl pentan-2-ylmalonate,acmc-1brsl,diethyl sec-amyl malonate
95449
diethyl 2-pentan-2-ylpropanedioate
CCCC(C)C(C(=O)OCC)C(=O)OCC
117-47-5
C12H22O4
230.304
MFCD00051558
RQFSNEWORATSCC-UHFFFAOYSA-N
diethyl 1-methylbutyl malonate,diethyl 2-pentyl malonate,diethyl 1-methylbutylmalonate,diethyl 1-methylbutal malonate,1-methylbutyl malonic ester,1-methylbutyl malonic acid diethyl ester,diethyl pentan-2-ylmalonate,acmc-1brsl,diethyl sec-amyl malonate
95449
diethyl 2-pentan-2-ylpropanedioate
CCCC(C)C(C(=O)OCC)C(=O)OCC
Applications
Diethyl (2-pentyl)malonate is used in vitro reaction of barbiturates with formaldehyde to evaluate forensic toxicology on the cause of death.

Notes
Incompatible with oxidizing agents.
Refractive Index 1.427
Density 0.968
Quantity 250g
Boiling Point 118°C to 119°C (9mmHg)
Formula Weight 230.31
Percent Purity 94%
Chemical Name or Material Diethyl (2-pentyl)malonate

EINECSNumber : 204-193-4

TSCA : No

Recommended Storage : Ambient temperatures