Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl 2-thienylmethylphosphonate, 95%

Click to view available options
Quantity:
250mg
1g
5g
2026-42-8
C9H15O3PS
234.25
MFCD00128457
GOJBUVQDOFHBLF-UHFFFAOYSA-N
diethyl thiophen-2-ylmethyl phosphonate,diethyl 2-thienylmethyl phosphonate,thiophen-2-ylmethyl-phosphonic acid diethyl ester,diethyl 2-thienylmethylphosphonate,2-diethoxyphosphorylmethyl thiophene,diethyl thiophen-2-ylmethyl phosphonate,phosphonic acid, 2-thienylmethyl-, diethyl ester,diethyl thiophen-2-ylmethylphosphonate,ksc207o5j,2-diethoxyphosphinylmethyl thiophene
4185897
2-(diethoxyphosphorylmethyl)thiophene
CCOP(=O)(CC1=CC=CS1)OCC
2026-42-8
C9H15O3PS
234.25
MFCD00128457
GOJBUVQDOFHBLF-UHFFFAOYSA-N
diethyl thiophen-2-ylmethyl phosphonate,diethyl 2-thienylmethyl phosphonate,thiophen-2-ylmethyl-phosphonic acid diethyl ester,diethyl 2-thienylmethylphosphonate,2-diethoxyphosphorylmethyl thiophene,diethyl thiophen-2-ylmethyl phosphonate,phosphonic acid, 2-thienylmethyl-, diethyl ester,diethyl thiophen-2-ylmethylphosphonate,ksc207o5j,2-diethoxyphosphinylmethyl thiophene
4185897
2-(diethoxyphosphorylmethyl)thiophene
CCOP(=O)(CC1=CC=CS1)OCC
Refractive Index 1.5200
Quantity 250mg
Color Yellow
Beilstein 1683411
Formula Weight 234.25
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Diethyl 2-thienylmethylphosphonate