Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (2S,3S)-(+)-2,3-epoxysuccinate, 98%, ACROS Organics™

Click to view available options
Quantity:
100mg
250mg
Packaging:
Glass Bottle in Plastic Container
73890-18-3
C8H12O5
188.179
MFCD01632600
LDFQMMUIJQDSAB-WDSKDSINSA-N
2s,3s-diethyl oxirane-2,3-dicarboxylate,diethyl 2s,3s-oxirane-2,3-dicarboxylate,2,3-diethyl 2s,3s-oxirane-2,3-dicarboxylate,2s,3s-2,3-oxiranedicarboxylic acid, diethyl ester,zlchem 164,2,3-oxiranedicarboxylicacid, 2,3-diethyl ester, 2s,3s,chembl88967,diethyl 2s-trans-oxiranedicarboxylate,2s,3s-epoxybutanedioic acid diethyl ester,2s,3s-2,3-epoxybutanedioic acid diethyl ester
7021072
diethyl (2S,3S)-oxirane-2,3-dicarboxylate
CCOC(=O)C1C(O1)C(=O)OCC
73890-18-3
C8H12O5
188.179
MFCD01632600
LDFQMMUIJQDSAB-WDSKDSINSA-N
2s,3s-diethyl oxirane-2,3-dicarboxylate,diethyl 2s,3s-oxirane-2,3-dicarboxylate,2,3-diethyl 2s,3s-oxirane-2,3-dicarboxylate,2s,3s-2,3-oxiranedicarboxylic acid, diethyl ester,zlchem 164,2,3-oxiranedicarboxylicacid, 2,3-diethyl ester, 2s,3s,chembl88967,diethyl 2s-trans-oxiranedicarboxylate,2s,3s-epoxybutanedioic acid diethyl ester,2s,3s-2,3-epoxybutanedioic acid diethyl ester
7021072
diethyl (2S,3S)-oxirane-2,3-dicarboxylate
CCOC(=O)C1C(O1)C(=O)OCC
Specific Rotation Condition + 108.00 (20.00°C c=2,diethylether)
Packaging Glass Bottle in Plastic Container
Quantity 250mg
Infrared Spectrum Authentic
Specific Rotation +104° to +112° (20°C, 589nm) (c=2, diethylether)
Refractive Index 1.4370 to 1.4390 (20°C, 589nm)
Color Colorless to Yellow
Assay Percent Range 98%
Formula Weight 188.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Diethyl (2S, 3S)-(+)-2, 3-epoxysuccinate, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.