Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl (3-aminopropyl)phosphonate, 95%

Manufacturer:  Alfa Aesar™ H51874MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51874MD

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
Diethyl (3-aminopropyl)phosphonate is used in the preparation of tert-Butyl (2-((diethoxyphosphoryl)propyl)amino)-3,4-dioxocyclobut-1- en-1-yl)carbamate by reacting with tert-butyl (2-ethoxy-3,4-dioxocyclobut-1-en-1-yl)carbamate.

Notes
Air sensitive. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

4402-24-8
195.2
UN2735
Diethyl (3-aminopropyl)phosphonate
C7H18NO3P
MFCD11110596
(3-Aminopropyl)phosphonic acid diethyl ester

Specifications

250mg
C7H18NO3P
MFCD11110596
(3-Aminopropyl)phosphonic acid diethyl ester
95%
4402-24-8
195.2
UN2735
Diethyl (3-aminopropyl)phosphonate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit