Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 4,4′-Azodibenzoate 95.0+%, TCI America™

Click to view available options
Quantity:
1g
7250-68-2
C18H18N2O4
326.352
MFCD00096305
YFFNOBALALSBCH-UHFFFAOYSA-N
4,4′C-Azodibenzoic Acid Diethyl Ester, Diethyl Azobenzene-4,4′C-dicarboxylate, Azobenzene-4,4′C-dicarboxylic Acid Diethyl Ester
232541
ethyl 4-[(4-ethoxycarbonylphenyl)diazenyl]benzoate
CCOC(=O)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)OCC
7250-68-2
C18H18N2O4
326.352
MFCD00096305
YFFNOBALALSBCH-UHFFFAOYSA-N
4,4′C-Azodibenzoic Acid Diethyl Ester, Diethyl Azobenzene-4,4′C-dicarboxylate, Azobenzene-4,4′C-dicarboxylic Acid Diethyl Ester
232541
ethyl 4-[(4-ethoxycarbonylphenyl)diazenyl]benzoate
CCOC(=O)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)OCC
Color Undesignated
Melting Point 144°C
Quantity 1g
Formula Weight 326.35
Physical Form Crystal-Powder at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material Diethyl 4,4′-Azodibenzoate