Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (4-aminobutyl)phosphonate oxalate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
296775-92-3
C10H22NO7P
299.26
MFCD00800258
YRLZCLQUARLFRX-UHFFFAOYSA-N
diethyl 4-aminobutyl phosphonate oxalate,diethyl aminobutyl phosphonate oxalate,oxalic acid diethyl 4-aminobutylphosphonate,diethyl 4-aminobutyl phosphonate oxalate salt,4-diethoxyphosphorylbutan-1-amine; oxalic acid,4-aminobutyl-phosphonic acid diethyl ester oxalate salt
2733598
4-diethoxyphosphorylbutan-1-amine;oxalic acid
CCOP(=O)(CCCCN)OCC.C(=O)(C(=O)O)O
296775-92-3
C10H22NO7P
299.26
MFCD00800258
YRLZCLQUARLFRX-UHFFFAOYSA-N
diethyl 4-aminobutyl phosphonate oxalate,diethyl aminobutyl phosphonate oxalate,oxalic acid diethyl 4-aminobutylphosphonate,diethyl 4-aminobutyl phosphonate oxalate salt,4-diethoxyphosphorylbutan-1-amine; oxalic acid,4-aminobutyl-phosphonic acid diethyl ester oxalate salt
2733598
4-diethoxyphosphorylbutan-1-amine;oxalic acid
CCOP(=O)(CCCCN)OCC.C(=O)(C(=O)O)O
For C-C Bond Formation (Olefination)
Quantity 1g
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Melting Point 148°C to 154°C
Assay Percent Range 97%
Formula Weight 299.26
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material Diethyl (4-aminobutyl)phosphonate oxalate

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC