Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl acetonylsuccinate, 95%

Click to view available options
Quantity:
5g
25g
100g
1187-74-2
C11H18O5
230.26
MFCD00071579
AAPVOVKOHNCXJA-UHFFFAOYSA-N
diethyl 2-2-oxopropyl succinate,diethyl acetonylsuccinate,acetonylsuccinic acid diethyl ester,acetonyl succinic acid diethyl ester,butanedioic acid, 2-oxopropyl-, diethyl ester,1,4-diethyl 2-2-oxopropyl butanedioate,acetonyl succinic,acetonylsuccinic aci,diethyl 2-acetonylbutanedioate,diethyl 2-2-oxopropyl succite
5463816
diethyl 2-(2-oxopropyl)butanedioate
CCOC(=O)CC(CC(=O)C)C(=O)OCC
1187-74-2
C11H18O5
230.26
MFCD00071579
AAPVOVKOHNCXJA-UHFFFAOYSA-N
diethyl 2-2-oxopropyl succinate,diethyl acetonylsuccinate,acetonylsuccinic acid diethyl ester,acetonyl succinic acid diethyl ester,butanedioic acid, 2-oxopropyl-, diethyl ester,1,4-diethyl 2-2-oxopropyl butanedioate,acetonyl succinic,acetonylsuccinic aci,diethyl 2-acetonylbutanedioate,diethyl 2-2-oxopropyl succite
5463816
diethyl 2-(2-oxopropyl)butanedioate
CCOC(=O)CC(CC(=O)C)C(=O)OCC
Refractive Index 1.4410
Quantity 100g
Boiling Point 135°C (1.3mmHg)
Density 1.074g/mL
Color Colorless to Yellow
Beilstein 1911511
Formula Weight 230.26
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Diethyl acetonylsuccinate