Alfa Aesar™ Diethyl acetylmethylphosphonate, 97%

Manufacturer: Alfa Aesar™ H2689603

Quantity 1g

 View more versions of this product

Catalog No. AAH2689603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1067-71-6
    Molecular Formula C7H15O4P
    Formula Weight 194.17
    MDL Number MFCD00044728
    Synonym Diethyl (2-oxopropyl)phosphonate
    Chemical Name or Material Diethyl acetylmethylphosphonate

    Specifications

    Formula Weight 194.17
    Flash Point 113°C (235°F)
    Quantity 1g
    Molecular Formula C7H15O4P
    MDL Number MFCD00044728
    CAS 1067-71-6
    Boiling Point 129°C (9mmHg)
    Synonym Diethyl (2-oxopropyl)phosphonate
    Chemical Name or Material Diethyl acetylmethylphosphonate
    Assay 97%

    Description

    Applications
    Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

    Solubility
    Miscible with tetrahydrofuran, ether, dichloromethane and chloroform.

    Notes
    Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit