Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Acetylphosphonate 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
919-19-7
C6H13O4P
180.14
MFCD00015138
YOHJPFQGGNEGSE-UHFFFAOYSA-N
Acetylphosphonic Acid Diethyl Ester
522997
1-diethoxyphosphorylethanone
CCOP(=O)(C(=O)C)OCC
919-19-7
C6H13O4P
180.14
MFCD00015138
YOHJPFQGGNEGSE-UHFFFAOYSA-N
Acetylphosphonic Acid Diethyl Ester
522997
1-diethoxyphosphorylethanone
CCOP(=O)(C(=O)C)OCC
Quantity 5g
Boiling Point 115°C
Color Yellow
Formula Weight 180.14
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material Diethyl Acetylphosphonate