Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl allylphosphonate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1067-87-4
C7H15O3P
178.17
MFCD00015134
YPJHXRAHMUKXAE-UHFFFAOYSA-N
diethyl allylphosphonate,diethyl prop-2-en-1-ylphosphonate,diethyl-allylphosphonate,acmc-1bprn,phosphonic acid, 2-propenyl-, diethyl ester,allylphosphonic acid diethyl,3-diethoxyphosphoryl-prop-1-ene,allylphosphonic acid diethyl ester,allyl phosphonic acid diethyl ester
573014
3-diethoxyphosphorylprop-1-ene
CCOP(=O)(CC=C)OCC
1067-87-4
C7H15O3P
178.17
MFCD00015134
YPJHXRAHMUKXAE-UHFFFAOYSA-N
diethyl allylphosphonate,diethyl prop-2-en-1-ylphosphonate,diethyl-allylphosphonate,acmc-1bprn,phosphonic acid, 2-propenyl-, diethyl ester,allylphosphonic acid diethyl,3-diethoxyphosphoryl-prop-1-ene,allylphosphonic acid diethyl ester,allyl phosphonic acid diethyl ester
573014
3-diethoxyphosphorylprop-1-ene
CCOP(=O)(CC=C)OCC
For C-C Bond Formation (Olefination)
Quantity 5g
Boiling Point 97.0°C
Packaging Glass bottle
Color Colorless
Refractive Index 1.4325 to 1.4345
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula (O)P(CH3CH2O)2CH2CHCH2
Formula Weight 178.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Diethyl allylphosphonate, 97%
Show More Show Less