Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl aminomethylenemalonate, 98%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
6296-99-7
C8H13NO4
187.195
MFCD01075695
WDTWMEZMHUEZKH-UHFFFAOYSA-N
diethyl 2-aminomethylene malonate, diethyl aminomethylenemalonate, 1,3-diethyl 2-aminomethylidene propanedioate, propanedioic acid, aminomethylene-, diethyl ester, diethyl aminomethylene malonate, amino-methylene malonic acid, diethyl ester, acmc-1b66v, ksc494c4d, diethyl amino-methylenemalonate
226751
diethyl 2-(aminomethylidene)propanedioate
CCOC(=O)C(=CN)C(=O)OCC
6296-99-7
C8H13NO4
187.195
MFCD01075695
WDTWMEZMHUEZKH-UHFFFAOYSA-N
diethyl 2-aminomethylene malonate, diethyl aminomethylenemalonate, 1,3-diethyl 2-aminomethylidene propanedioate, propanedioic acid, aminomethylene-, diethyl ester, diethyl aminomethylene malonate, amino-methylene malonic acid, diethyl ester, acmc-1b66v, ksc494c4d, diethyl amino-methylenemalonate
226751
diethyl 2-(aminomethylidene)propanedioate
CCOC(=O)C(=CN)C(=O)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 60°C to 65°C
Boiling Point 114°C (1mmHg)
Beilstein 1708651
Quantity 5g
Formula Weight 187.2
Percent Purity 98%
Chemical Name or Material Diethyl aminomethylenemalonate

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c

H315-H317-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only