Alfa Aesar™ Diethyl bis(2-cyanoethyl)malonate, 98+%

Manufacturer: Alfa Aesar™ B23277-06

Quantity 5g

 View more versions of this product

Catalog No. AAB2327706

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 1444-05-9
    Molecular Formula C13H18N2O4
    Formula Weight 266.30g/mol
    MDL Number MFCD00013831
    UN Number UN3439
    Beilstein 1800053
    Synonym Bis(2-cyanoethyl)malonic acid diethyl ester; Bis(4,4-ethoxycarbonyl)pimelonitrile
    Chemical Name or Material Diethyl bis(2-cyanoethyl)malonate

    Specifications

    Boiling Point 174° to 176°C (0.5mmHg)
    Melting Point 60° to 63°C
    Quantity 5g
    CAS 1444-05-9
    Molecular Formula C13H18N2O4
    Formula Weight 266.30g/mol
    MDL Number MFCD00013831
    UN Number UN3439
    Beilstein 1800053
    Synonym Bis(2-cyanoethyl)malonic acid diethyl ester; Bis(4,4-ethoxycarbonyl)pimelonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H312-H332
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    TSCA TSCA
    Chemical Name or Material Diethyl bis(2-cyanoethyl)malonate
    Assay ≥98%

    GHS H Statement
    H301-H312-H332
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit