Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl bis(2-cyanoethyl)malonate, 98+%

Manufacturer:  Alfa Aesar™ B2327714

Quantity 25g

 View more versions of this product

Catalog No. AAB2327714


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

1444-05-9
266.3
UN3439
Bis(2-cyanoethyl)malonic acid diethyl ester; Bis(4,4-ethoxycarbonyl)pimelonitrile
C13H18N2O4
MFCD00013831
1800053
Diethyl bis(2-cyanoethyl)malonate

Specifications

25g
C13H18N2O4
MFCD00013831
174° to 176°C (0.5mmHg)
60° to 63°C
Diethyl bis(2-cyanoethyl)malonate
1444-05-9
266.3
UN3439
1800053
Bis(2-cyanoethyl)malonic acid diethyl ester; Bis(4,4-ethoxycarbonyl)pimelonitrile
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit