Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl carbonate, 99%, anhydrous, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
1L
105-58-8
C5H10O3
118.13
OIFBSDVPJOWBCH-UHFFFAOYSA-N
ethyl carbonate,carbonic acid, diethyl ester,carbonic acid diethyl ester,eufin,diatol,diaethylcarbonat,carbonic ether,ethoxyformic anhydride,diethylkarbonat,ethyl carbonate eto 2co
7766
diethyl carbonate
CCOC(=O)OCC
105-58-8
C5H10O3
118.13
OIFBSDVPJOWBCH-UHFFFAOYSA-N
ethyl carbonate,carbonic acid, diethyl ester,carbonic acid diethyl ester,eufin,diatol,diaethylcarbonat,carbonic ether,ethoxyformic anhydride,diethylkarbonat,ethyl carbonate eto 2co
7766
diethyl carbonate
CCOC(=O)OCC
Packaging AcroSeal™ Glass Bottle
Viscosity 0.827 mPa.s (20°C)
Boiling Point 126.0°C to 128.0°C
Quantity 1L
Water 0.1% max.
Melting Point -43.0°C
Flash Point 33°C
Density 0.9750g/mL
Infrared Spectrum Authentic
Refractive Index 1.3830 to 1.3850
Specific Gravity 0.975
Assay Percent Range 98.5% min. (GC)
Linear Formula (C2H5O)2CO
Merck Index 15,3835
Solubility Information Solubility in water: 18.8g/L.
Formula Weight 118.13
Percent Purity 99%
Chemical Name or Material Diethyl carbonate
Show More Show Less

GHS H Statement Flammable liquid and vapor.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS Signal Word: Warning

EINECSNumber : 203-311-1