Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Diethyl D-tartrate, 99%, made from unnatural tartaric acid, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
500mL
Packaging:
Glass bottle
13811-71-7
C8H14O6
206.19
MFCD00064451
YSAVZVORKRDODB-WDSKDSINSA-N
--diethyl d-tartrate,2s,3s-diethyl 2,3-dihydroxysuccinate,diethyl d-tartrate,d---tartaric acid diethyl ester,diethyl d---tartrate,diethyl-d-tartrate,diethyl 2s,3s-2,3-dihydroxybutanedioate,2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester,--diethyl-d-tartrate,diethyl s-r*,r*-tartrate
117410
diethyl (2S,3S)-2,3-dihydroxybutanedioate
CCOC(=O)C(C(C(=O)OCC)O)O
13811-71-7
C8H14O6
206.19
MFCD00064451
YSAVZVORKRDODB-WDSKDSINSA-N
--diethyl d-tartrate,2s,3s-diethyl 2,3-dihydroxysuccinate,diethyl d-tartrate,d---tartaric acid diethyl ester,diethyl d---tartrate,diethyl-d-tartrate,diethyl 2s,3s-2,3-dihydroxybutanedioate,2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester,--diethyl-d-tartrate,diethyl s-r*,r*-tartrate
117410
diethyl (2S,3S)-2,3-dihydroxybutanedioate
CCOC(=O)C(C(C(=O)OCC)O)O
Specific Gravity 1.2
Melting Point 17.0°C
Packaging Glass bottle
Flash Point 93°C
Color Colorless
Quantity 500mL
Refractive Index 1.4450 to 1.4470
Boiling Point 280.0°C
Specific Rotation Condition −9° (20°C neat)
Specific Rotation − 9.00
Infrared Spectrum Authentic
Density 1.2000g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula [-CH(OH)CO2C2H5]2
Beilstein 03, I, 181
Fieser 11,181; 13,202; 16,54
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Formula Weight 206.19
Physical Form Viscous Liquid
Percent Purity 99%
Chemical Name or Material (-)-Diethyl D-tartrate, Made from unnatural tartaric acid
Show More Show Less

EINECSNumber : 237-458-8