Alfa Aesar™ Diethyl di-n-propylmalonate, 97%

Manufacturer: Alfa Aesar™ B2506236

Quantity 500g

 View more versions of this product

Catalog No. AAB2506236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6065-63-0
    Molecular Formula C13H24O4
    Formula Weight 244.33
    MDL Number MFCD00026840
    Synonym Di-n-propylmalonic acid diethyl ester
    Chemical Name or Material Diethyl di-n-propylmalonate

    Specifications

    Formula Weight 244.33
    Molecular Formula C13H24O4
    MDL Number MFCD00026840
    CAS 6065-63-0
    Synonym Di-n-propylmalonic acid diethyl ester
    Quantity 500g
    Chemical Name or Material Diethyl di-n-propylmalonate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit