Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl di-n-propylmalonate, 97%

Manufacturer:  Alfa Aesar™ B2506222

Quantity 100g

 View more versions of this product

Catalog No. AAB2506222


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

6065-63-0
244.33
Di-n-propylmalonic acid diethyl ester
C13H24O4
MFCD00026840
Diethyl di-n-propylmalonate

Specifications

100g
C13H24O4
MFCD00026840
Diethyl di-n-propylmalonate
6065-63-0
244.33
Di-n-propylmalonic acid diethyl ester
97%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit