Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl di-n-propylmalonate, 97%

Click to view available options
Quantity:
25g
100g
500g
6065-63-0
C13H24O4
244.331
MFCD00026840
NNDOHYGFLASMFR-UHFFFAOYSA-N
diethyl dipropylmalonate,diethyl 2,2-dipropylmalonate,diethyl-dipropylmalonate,diethyldipropylmalonate,diethyl di-n-propylmalonate,1,3-diethyl 2,2-dipropylpropanedioate,propanedioic acid, dipropyl-, diethyl ester,malonic acid, dipropyl-, diethyl ester,acmc-209mla,diethyl2,2-dipropylmalonate
80168
diethyl 2,2-dipropylpropanedioate
CCCC(CCC)(C(=O)OCC)C(=O)OCC
6065-63-0
C13H24O4
244.331
MFCD00026840
NNDOHYGFLASMFR-UHFFFAOYSA-N
diethyl dipropylmalonate,diethyl 2,2-dipropylmalonate,diethyl-dipropylmalonate,diethyldipropylmalonate,diethyl di-n-propylmalonate,1,3-diethyl 2,2-dipropylpropanedioate,propanedioic acid, dipropyl-, diethyl ester,malonic acid, dipropyl-, diethyl ester,acmc-209mla,diethyl2,2-dipropylmalonate
80168
diethyl 2,2-dipropylpropanedioate
CCCC(CCC)(C(=O)OCC)C(=O)OCC
Quantity 25g
Formula Weight 244.33
Percent Purity 97%
Chemical Name or Material Diethyl di-n-propylmalonate