Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl diethylmalonate, 98%

Click to view available options
Quantity:
25g
100g
500g
77-25-8
C11H20O4
216.277
MFCD00009132
ZKBBUZRGPULIRN-UHFFFAOYSA-N
diethyl diethylmalonate,diethyl 2,2-diethylmalonate,diethylmalonic ester,propanedioic acid, diethyl-, diethyl ester,diethylmalonate diethyl ester,diethylmalonic acid diethyl ester,diethylmalonic acid, diethyl ester,unii-ceh13944yq,1,3-diethyl 2,2-diethylpropanedioate,diethyl diethylmalonate diethyl
66165
diethyl 2,2-diethylpropanedioate
CCC(CC)(C(=O)OCC)C(=O)OCC
77-25-8
C11H20O4
216.277
MFCD00009132
ZKBBUZRGPULIRN-UHFFFAOYSA-N
diethyl diethylmalonate,diethyl 2,2-diethylmalonate,diethylmalonic ester,propanedioic acid, diethyl-, diethyl ester,diethylmalonate diethyl ester,diethylmalonic acid diethyl ester,diethylmalonic acid, diethyl ester,unii-ceh13944yq,1,3-diethyl 2,2-diethylpropanedioate,diethyl diethylmalonate diethyl
66165
diethyl 2,2-diethylpropanedioate
CCC(CC)(C(=O)OCC)C(=O)OCC
Quantity 25g
Flash Point 94°C (202°F)
Boiling Point 228°C to 230°C
Merck Index 14,3789
Formula Weight 216.28
Percent Purity 98%
Chemical Name or Material Diethyl diethylmalonate