Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, 99%, Alfa Aesar™

Click to view available options
Quantity:
1L
4L
4 x 1L
4 x 4L
60-29-7
C4H10O
74.123
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
60-29-7
C4H10O
74.123
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
UN Number UN1155
Formula Weight 74.12
Physical Form Liquid
Percent Purity 99%
Grade HPLC
Packaging Glass bottle
Specific Gravity 0.713
Flash Point −45°C
Boiling Point 34.6°C
Melting Point -116°C
Quantity 4 x 4L
Chemical Name or Material Diethyl ether
Show More Show Less