Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, 99%, Alfa Aesar™

Click to view available options
Quantity:
1L
4 x 1L
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
Assay Percent Range 99%
Solubility Information Immiscible with water,miscible with alcohol.
Formula Weight 74.12
Physical Form Liquid
Grade HPLC
Density 0.713 g/mL
Flash Point -45°C (-49°F)
Boiling Point 34.6°C
Melting Point °C to 116°C
Quantity 1L
Chemical Name or Material Diethyl ether
Show More Show Less

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c

H224-H302-H305-H335-H336-H373

DOTInformation : Hazard Class: 3; Packaging Group: I

EINECSNumber : 200-467-2

RTECSNumber : KI5775000

TSCA : Yes

Recommended Storage : Ambient temperatures