Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 38990K7

Quantity 4L

 View more versions of this product

Catalog No. AA38990K7

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

60-29-7
99%
74.12
Diethyl ether
Liquid
(C2H5)2O
MFCD00011646

Specifications

60-29-7
99%
74.12
0.713g/mL
Diethyl ether
Liquid
(C2H5)2O
MFCD00011646
4L

Safety and Handling

SDS & Safety Documents

EINECSNumber : 200-467-2

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit