Diethyl ether, >99%, inhibitor free, Alfa Aesar™

Solvent For U.V. Spectrophotometric and General Use

Manufacturer: Alfa Aesar 40976K7

Quantity 4L

 View more versions of this product

Catalog No. AA40976K7

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 60-29-7
    Physical Form Liquid
    Assay Percent Range >99%
    Molecular Formula C4H10O
    Formula Weight 74.12
    MDL Number MFCD00011646
    Synonym Ether
    Chemical Name or Material Diethyl ether
    Grade Spectrophotometric grade

    Specifications

    Boiling Point 34.6°C
    Physical Form Liquid
    Color Clear colorless
    Melting Point −116°C
    Formula Weight 74.12
    Quantity 4L
    DOT Information DOT Class: 3, Packaging Group: I
    Flash Point −45°C
    UN Number 1155
    Density 0.713g/cm3 at 20°C
    EINECS Number 200-467-2
    Ignition Point 170°C (338°F)
    Odor Sweet
    Vapor Pressure 586.3 hPa (440mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range >99%
    Molecular Formula C4H10O
    MDL Number MFCD00011646
    CAS 60-29-7
    Synonym Ether
    Chemical Name or Material Diethyl ether
    Grade Spectrophotometric grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit